Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN450plus, 15 mm
Product Code. 17518655 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN450plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17518655 SN450plus Sterilizer Each
1 options
Product Code. 17518655 Supplier Memmert™ Supplier No. SN450plusB02075
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SN450plus Sterilizer
Product Type Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel