Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN75, 15 mm
Product Code. 17518765 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN75 Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17518765 SN75 Sterilizer Each
1 options
Product Code. 17518765 Supplier Memmert™ Supplier No. SN75B02072
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SN75 Sterilizer
Product Type Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel