Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
Specifications
| For Use With (Equipment) | SN750plus Sterilizer |
| Product Type | Drip Tray |
| Height (Metric) Tray | Rim: 15 mm |
| Material | Stainless Steel |