missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 100mm Dia., in Millimeter Pore Sizes with ASTM E 11

Brand:  RETSCH 60.108.031500

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 10044871

  • 150.00 € / Each

Estimated Shipment: 21-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1 Each
Test Sieve
Shaker AS 200
40mm
For separation, fractioning, particle size determination
100mm
31.5mm
ASTM E 11
Certificate indicating Compliance
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers