missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50mmH, with Perforated Plates with Square Holes
Brand: RETSCH 60.190.125000
Additional Details : Weight : 1.00000kg
Specifications
Stainless Steel | |
200mm | |
1 Each | |
Test Sieve | |
Certificate indicating Compliance |
Stainless Steel | |
For separation, fractioning, particle size determination | |
125mm | |
ISO 3310/2 | |
50mm |