missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 400mm Dia., with Perforated Plates with Round Holes

Brand:  RETSCH 60.162.005600

Code : A0

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 10012692

  • 393.00 € / Each

Estimated Shipment: 23-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
400mm
1 Each
Test Sieve
Certificate indicating Compliance
Stainless Steel
For separation, fractioning, particle size determination
5.6mm
ISO 3310/2
65mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers