Learn More
RETSCH Stainless-Steel Test Sieves, 400mm Dia., with Perforated Plates with Round Holes
381.00 € - 670.00 €
Specifications
Diameter (Metric) Outer | 400 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1 Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10307902
|
RETSCH
60.162.001250 |
1.25mm |
670.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10368372
|
RETSCH
60.162.106000 |
106mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10154732
|
RETSCH
60.162.100000 |
100mm |
393.00 €
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
10572863
|
RETSCH
60.162.010000 |
10mm |
381.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10798784
|
RETSCH
60.162.011200 |
11.2mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10378372
|
RETSCH
60.162.112000 |
112mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10675163
|
RETSCH
60.162.012500 |
12.5mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10164732
|
RETSCH
60.162.125000 |
125mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10124542
|
RETSCH
60.162.013200 |
13.2mm |
393.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10092792
|
RETSCH
60.162.016000 |
16mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10685163
|
RETSCH
60.162.014000 |
14mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10645743
|
RETSCH
60.162.018000 |
18mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10032822
|
RETSCH
60.162.019000 |
19mm |
393.00 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10462262
|
RETSCH
60.162.002360 |
2.36mm |
650.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10276652
|
RETSCH
60.162.001000 |
1mm |
670.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10338512
|
RETSCH
60.162.002500 |
2.5mm |
670.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10286362
|
RETSCH
60.162.020000 |
20mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10296262
|
RETSCH
60.162.002800 |
2.8mm |
670.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10104782
|
RETSCH
60.162.022400 |
22.4mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10412272
|
RETSCH
60.162.025000 |
25mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10412462
|
RETSCH
60.162.026500 |
26.5mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10686123
|
RETSCH
60.162.028000 |
28mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10183812
|
RETSCH
60.162.002000 |
2mm |
670.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10739774
|
RETSCH
60.162.003150 |
3.15mm |
650.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10236412
|
RETSCH
60.162.003350 |
3.35mm |
670.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10032832
|
RETSCH
60.162.003550 |
3.55mm |
670.00 €
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
10473032
|
RETSCH
60.162.037500 |
37.5mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10626703
|
RETSCH
60.162.004500 |
4.5mm |
381.00 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10738794
|
RETSCH
60.162.031500 |
31.5mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10799944
|
RETSCH
60.162.045000 |
45mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10105022
|
RETSCH
60.162.040000 |
40mm |
393.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
10789934
|
RETSCH
60.162.004750 |
4.75mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10574593
|
RETSCH
60.162.035500 |
35.5mm |
381.00 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10625933
|
RETSCH
60.162.004000 |
4mm |
381.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
10012692
|
RETSCH
60.162.005600 |
5.6mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10206802
|
RETSCH
60.162.053000 |
53mm |
393.00 €
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
10338182
|
RETSCH
60.162.056000 |
56mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10002692
|
RETSCH
60.162.005000 |
5mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10012732
|
RETSCH
60.162.050000 |
50mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10464182
|
RETSCH
60.162.006300 |
6.3mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10474182
|
RETSCH
60.162.006700 |
6.7mm |
381.00 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
10012742
|
RETSCH
60.162.063000 |
63mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10348132
|
RETSCH
60.162.007100 |
7.1mm |
393.00 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
10236322
|
RETSCH
60.162.071000 |
71mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10296362
|
RETSCH
60.162.075000 |
75mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10296742
|
RETSCH
60.162.008000 |
8mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10328232
|
RETSCH
60.162.080000 |
80mm |
393.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10494182
|
RETSCH
60.162.090000 |
90mm |
432.00 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10634593
|
RETSCH
60.162.009500 |
9.5mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10768624
|
RETSCH
60.162.009000 |
9mm |
381.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Specifications
400 mm | |
Stainless Steel | |
1 Each | |
ISO 3310/2 | |
65mm |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Certificate indicating Compliance |