missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 400mm Dia., with Stainless-Steel Wire

Brand:  RETSCH 60.166.014000

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 10024911

  • 428.00 € / Each

Estimated Shipment: 30-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
400mm
1 Each
Test Sieve
Vibratory Shaker AS 450
65mm
Stainless Steel
For separation, fractioning, particle size determination
14mm
ISO 3310/1
Certificate indicating Compliance
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers