missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 400mm Dia., with Stainless-Steel Wire

Brand:  RETSCH 60.166.008000

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 10045051

  • 360.00 € / Each

Estimated Shipment: 25-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1 Each
Test Sieve
Vibratory Shaker AS 450
65mm
For separation, fractioning, particle size determination
400mm
8mm
ISO 3310/1
Certificate indicating Compliance
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers