Learn More
Thermo Scientific Chemicals Stains-All, 96%
Description
General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Formula Weight | 559.58g/mol |
| Beilstein | 3865213 |
| Product Type | Stain |
| Quantity | 1g |
| Assay | 0.96 |
Frequently Asked Questions (FAQs)
Yes, Stains-All, 96% is a cationic cyanine dye, and it is possible to stain RNA with it. Gel staining should be performed in the dark, as cyanine dyes can be excited with visible light to produce ROS, leading to damage of biomolecules.
Safety and Handling
- Stains-All
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: 1-ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazol-2(1H)-ylidene)-2-methyl-1-propenyl]-naphtho[1,2-d]thiazol
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.