missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Stains-all, 97%, pure

Product Code. 10276840 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10276840

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7423-31-6 | C30H28BrN2S2+ | 560.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-31-6
Molecular Formula C30H28BrN2S2+
Molecular Weight (g/mol) 560.592
MDL Number MFCD00078788
InChI Key MPBRYMWMMKKRGC-UHFFFAOYSA-N
Synonym 1-Ethyl-2-[3-(1-ethylnaphtho[1, 2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1, 2-d]thiazolium bromide
PubChem CID 123134119
IUPAC Name (2E)-1-ethyl-2-[(E)-3-(1-ethylbenzo[e][1,3]benzothiazol-1-ium-2-yl)-2-methylprop-2-enylidene]benzo[e][1,3]benzothiazole;hydrobromide
SMILES CCN1C(=CC(=CC2=[N+](C3=C(S2)C=CC4=CC=CC=C43)CC)C)SC5=C1C6=CC=CC=C6C=C5.Br

Specifications

Assay Percent Range 97%
Beilstein 27, IV, 8619
Infrared Spectrum Authentic
Formula Weight 559.59
Percent Purity 97%
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Gray to Purple
Quantity 1 g
Physical Form Powder
Chemical Name or Material Stains-all, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.