missing translation for 'onlineSavingsMsg'
Learn More

Standard 1506: Ca @ 2000 μg/g; P, Zn, Mo @ 800 μg/g in 75 cSt Oil

Product Code. 30322585
Change view
Click to view available options
Quantity:
400 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30322585 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322585 Supplier VHG Labs Supplier No. VHGJET1506400G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium∣Molybdenum∣Phosphorus∣Zinc
CAS 7440-70-2, 7439-98-7, 7723-14-0, 7440-66-6
Analyte or Component Names Calcium∣Molybdenum∣Phosphorus∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix Hydrocarbon Oil
Industry Type Industrial QC
Quantity 400 g
Concentration or Composition (by Analyte or Components) Molybdenum 800 μg/g∣Phosphorus 800 μg/g∣Zinc 800 μg/g∣Calcium 2000 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.