missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Solution, SI Analytics™

Product Code. 10035210 Shop All SI Analytics Products
Click to view available options
Quantity:
60 Ampules
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10035210

Brand: SI Analytics 285138262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SI Analytics™ Standard Buffer Solution pH 9.18 resists changes in pH when small quantities of an alkali are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, disodium tetraborate decahydrate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Colorless
Density 1g/cm3
pH 9.18
Physical Form Liquid
Quantity 60 Ampules
Formulation Water, disodium tetraborate decahydrate
Odor Odorless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.