Learn More
Buffer Solution, pH 4.001, Reagecon™
Certified by an ISO 17025 accredited test method and NIST traceable. Buffer Solution, pH 4.001, Reagecon™ is tested at 20°C to a standard NIST value. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.
Brand: Reagecon 104001
Code : A0
Additional Details : Weight : 0.54000kg
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Calibrated to a standard precision NIST value
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
0°C | |
Colorless | |
Liquid | |
pH Buffer | |
Yes |
100°C | |
4.001 | |
500 mL | |
Characteristic |