Learn More
Neogen Petrifilm™ Staph Express Count Plate
Description
- Results in 24 Hours.
- Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
- Unique system to facilitate the enumeration of S. aureus
- This plate has been awarded Official Methods of Analysis (OMA) #2003.07 for processed and prepared foods, #2003.8 for select dairy foods and #2003.11 for Select meat, seafood , poultry and NF VALIDATION certificates: Neogen-01/9-04/03 A and Neogen-01/9-04/03 B for All human foods and Pet food by AFNOR Certification.
- Food processors and contract labs worldwide trust Neogen Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
| Product Type | Count Plate |
| Test Type | Food and Beverage, Environmental Test |
| Quantity | 500 |