missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05985-50ul
This item is not returnable.
View return policy
Description
STARD5 Polyclonal antibody specifically detects STARD5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| STARD5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| MGC10327, StARD5, StAR-related lipid transfer (START) domain containing 5, stAR-related lipid transfer protein 5, START domain containing 5, START domain-containing protein 5 | |
| Produced in rabbits immunized with E. coli-derived Human STARD5 fragment. | |
| 50 μg | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 80765 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction