Learn More
STAT3 Antibody (CL0490) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52935UV
Product Code. 30304471
  • 748.00 € / 0.10mL
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT3
Monoclonal
DyLight 350
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
This antibody was developed using a recombinant protein derived from P40763, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
6774
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL0490
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified