Learn More
STAT5A, p Tyr694 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82219
This item is not returnable.
View return policy
Description
STAT5A Polyclonal specifically detects STAT5A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Western Blot, Immunohistochemistry | |
| The antiserum was produced against synthesized phosphopeptide derived from human STAT5A around the phosphorylation site of tyrosine 694 (D-G-YP-V-K). | |
| 0.05 mL |