missing translation for 'onlineSavingsMsg'
Learn More
Learn More
State of Texas, C6-C35 Alkanes, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| n-Decane | 124-18-5 | 1000μg/mL |
| n-Dodecane | 112-40-3 | 1000μg/mL |
| n-Eicosane | 112-95-8 | 1000μg/mL |
| n-Heptadecane | 629-78-7 | 1000μg/mL |
| n-Heptane | 142-82-5 | 1000μg/mL |
| n-Hexadecane | 544-76-3 | 1000μg/mL |
| n-Hexane | 110-54-3 | 1000μg/mL |
| n-Nonadecane | 629-92-5 | 1000μg/mL |
| n-Nonane | 111-84-2 | 1000μg/mL |
| n-Octacosane | 630-02-4 | 1000μg/mL |
| n-Octadecane | 593-45-3 | 1000μg/mL |
| n-Octane | 111-65-9 | 1000μg/mL |
| n-Pentacosane | 629-99-2 | 1000μg/mL |
| n-Pentadecane | 629-62-9 | 1000μg/mL |
| n-Pentatriacontane | 630-07-9 | 1000μg/mL |
| n-Tetradecane | 629-59-4 | 1000μg/mL |
| n-Tridecane | 629-50-5 | 1000μg/mL |
| n-Undecane | 1120-21-4 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | State of Texas, C6-C35 Alkanes |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Pentane |
| Industry Type | Petrochemical Testing |
| Quantity | 1 mL |
Safety and Handling
Product Identifier
- State of Texas, C6-C35 Alkanes
- Danger
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : pentane, n-hexane
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction