Learn More
Melford Staurosporine
Descripción
Staurosporine is a potent natural compound and is a crucial tool in studying signal transduction pathways, cell cycle regulation, and cancer therapy.
Pharmacologists and pharmaceutical researchers leverage Staurosporine in drug discovery and development. Its ability to inhibit kinases makes it valuable in identifying potential therapeutic targets and assessing the efficacy of novel compounds for treating diseases like cancer. In cancer research, Staurosporine's role is pivotal. By interfering with key pathways involved in cell proliferation and survival, it provides insights into potential cancer treatments. Researchers explore its use as a chemotherapeutic agent, seeking ways to harness its properties for targeted cancer therapies.
Especificaciones
Especificaciones
| Chemical Name or Material | Staurosporine |
| CAS | 62996-74-1 |
| Quantity | 500 μg |
| Molecular Formula | C28H26N4O3 |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.