missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, 99+%, Thermo Scientific Chemicals

Product Code. 15495139
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495139 1 mg 1mg
15485139 5 mg 5mg
15405149 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Codice prodotto. 15495139 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' J62837.M^
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Questo articolo non è restituibile. Consulta la politica di reso

Staurosporine (staurosporin, CAS # 62996-74-1) is a bacterial alkyloid isolated from Streptomyces staurosporeus.

Broad spectrum protein kinase inhibitor Staurosporine is used as anti-fungal and anti-hypertensive, which results to study the anti-cancer treatment. It acts as a precursor to protein kinase inhibitor midostaurin. It is also used to induce cell death in Swiss Webster primary macrophage cells and to study the effect of caspase activation on human sperm quality.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Staurosporine is an indolocarbazole alkaloid capable of permeating cell membranes
  • Staurosporine has anti-cancer properties that can induce apoptosis
  • Staurosporine is a broad inhibitor of protein kinases, preventing growth and division cycles

Application

  • Staurosporine is used for research on acute myeloid leukemia
  • It is suitable for research applications including cell culture and biochemistry
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
MDL Number MFCD00077402
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym Antibiotic AM-2282
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifica

Type Staurosporine
Physical Form Powder
Melting Point 240°C to 243°C
Color Yellow
Quantity 1 mg
Beilstein 1060573
Merck Index 14,8802
Solubility Information Soluble in DMSO or ethanol;Soluble in dimethyl sulfoxide ,dimethyl formamide,ethyl acetate,hot acetone and ethanol; Slightly soluble in chloroform and methanol; Insoluble in water.
Formula Weight 466.54
Percent Purity ≥99%
Chemical Name or Material Staurosporine
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Staurosporine
Avvertenza
  • Warning
Categoria di pericolo
  • Carcinogenicity Category 2
Indicazioni di pericolo
  • H351-Suspected of causing cancer.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.