missing translation for 'onlineSavingsMsg'
Learn More

Stavudine, MedChemExpress

Product Code. 16478106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478106 100 mg 100mg
16432026 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478106 Supplier Medchem Express Supplier No. HYB0116
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Stavudine (d4T) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). Stavudine has activity against HIV-1 and HIV-2. Stavudine also inhibits the replication of mitochondrial DNA (mtDNA). Stavudine reduces NLRP3 inflammasome activation and modulates Amyloid-β autophagy. Stavudine induces apoptosis.

Stavudine (d4T) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). Stavudine has activity against HIV-1 and HIV-2. Stavudine also inhibits the replication of mitochondrial DNA (mtDNA). Stavudine reduces NLRP3 inflammasome activation and modulates Amyloid-β autophagy. Stavudine induces apoptosis.

Specifications

Chemical Name or Material Stavudine
Physical Form Solid
CAS 3056-17-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H12N2O4
For Use With (Application) COVID-19-anti-virus
Synonym d4T
Solubility Information DMSO : 125 mg/mL (557.51 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (74.35 mM; Need ultrasonic)
SMILES OC[C@H]1O[C@@H](N2C=C(C)C(NC2=O)=O)C=C1
Molecular Weight (g/mol) 224.21
Formula Weight 224.21
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.