missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STC2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148632
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
Specifications
| STC2 | |
| Polyclonal | |
| Unconjugated | |
| STC2 | |
| AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 20856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O88452 | |
| STC2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Stanniocalcin 2/STC-2, Thr25-Arg296. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction