missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STCH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80715
This item is not returnable.
View return policy
Description
STCH Polyclonal specifically detects STCH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| STCH | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| P48723 | |
| HSPA13 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDH | |
| 0.1 mL | |
| Signal Transduction | |
| 6782 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| heat shock 70 kDa protein 13, heat shock protein 70kDa family, member 13, microsomal stress 70 protein ATPase core, Microsomal stress-70 protein ATPase core, STCHStress-70 protein chaperone microsome-associated 60 kDa protein, stress 70 protein chaperone, microsome-associated, 60kD, stress 70 protein chaperone, microsome-associated, 60kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction