missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Stealth RNAi™ siRNA GAPDH Positive Control (human)

Product Code. 10206703
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10206703 250 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206703 Supplier Invitrogen™ Supplier No. 12935140
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stealth RNAi™ siRNA GAPDH Positive Control (human)

Stealth RNAi™ siRNA Positive Housekeeping Control Duplexes (human) are ideal for use in RNA interference (RNAi) experiments as a control for assessing knockdown and optimizing RNAi experiments. These controls are: –Designed to efficiently knock-down the intended target –Have been bench tested and are supplied in a ready-to-use format

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This individual control duplex is supplied as 250 μl at 20 μM and includes 1 ml of 1X RNA Annealing/Dilution Buffer. Store at -20°C. Products are guaranteed stable for 6 months when properly stored.
Control Type Positive Control
RNAi Type siRNA
For Use With (Application) RNAi
Label or Dye Unconjugated
Product Line Stealth RNAi™ siRNA
Product Type siRNA
Quantity 250 μL
Shipping Condition Dry Ice
Target GAPDH

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.