missing translation for 'onlineSavingsMsg'
Learn More

Stearalkonium Chloride-d7, TRC

Product Code. 30414692
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414692 10 mg 10mg
30414660 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30414692 Supplier Toronto Research Chemicals Supplier No. TRCS68570210MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27 H43 D7 N . Cl
Molecular Weight (g/mol) 431.19
Synonym N,N-Dimethyl-N-octadecylbenzenemethanaminium Chloride-d7, Benzyldimethyloctadecylammonium Chloride-d7, Algene SC 25-d7, Carsoquat SDQ 25-d7, Stebac-d7, Triton CG 500-d7, Varisoft SDC-d7
IUPAC Name N,N-Dimethyl-N-((phenyl-d5)methyl-d2)octadecan-1-aminium Chloride
SMILES [2H]C1=C(C([2H])=C(C([2H])=C1[2H])[2H])C([2H])([N+](C)(CCCCCCCCCCCCCCCCCC)C)[2H].[Cl-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H50N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-25-28(2,3)26-27-23-20-19-21-24-27;/h19-21,23-24H,4-18,22,25-26H2,1-3H3;1H/q+1;/p-1/i19D,20D,21D,23D,24D,26D2;
Formula Weight 430.4071
Chemical Name or Material Stearalkonium Chloride-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.