missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stella/Dppa3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41089-0.025mg
This item is not returnable.
View return policy
Description
Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Stella/Dppa3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DPPA3 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human Stella. The immunogen is located within the last 50 amino acids of Stella. Amino Acid Squence: NGWDPSENARIGN | |
| 0.025 mg | |
| Developmental Biology | |
| 359787 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction