Learn More
Description
For Complete Differentiation of Human Mesenchymal Stem Cells into Fat Cells
StemPro™ Adipogenesis Differentiation Kit has been developed for the adipogenic differentiation of mesenchymal stem cells (MSCs) in tissue culture vessels. The kit contains all reagents required for inducing MSCs to be committed to the adipogenesis pathway and generate adipocytes. Using StemPro™ Adipogenesis Differentiation Kit in combination with StemPro™ MSC SFM or MesenPRO RS™ Medium provides a standardized culture workflow solution for MSC isolation, expansion and differentiation into lipid vesicle-forming adipocytes.
Specifications
Specifications
| Cell Type | Stem Cells (Mesenchymal) |
| Content And Storage | This kit is shipped in two parts with separate storage requirements. These components are not sold individually: • StemPro™ Adipocyte Differentiation Basal Medium (100 ml; store in the dark at 2°C to 8°C) • StemPro™ Adipogenesis Supplement (10 ml; store in the dark at -5°C to -20°C) The Basal Medium ships at room temperature, while the Supplement ships on dry ice. |
| Culture Environment | CO2 |
| For Use With (Application) | Stem Cell Differentiation |
| Product Type | Adipogenesis Differentiation Kit |
| Endotoxin Level | ≤50 EU/mL, ≤ 50 EU/ml |
| Format | Liquid |
| Glucose | High Glucose |
| Glutamine | GlutaMAX™-I |
| HEPES Buffer | No HEPES |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.