Learn More
Thermo Scientific™ Sterilin™ Black and White Microtiter™ Plates
Product Code. 11359163
Change view
Click to view available options
Color:
Black
White
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11359163 Black Pack of 50
11718194 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11359163 Supplier Thermo Scientific™ Supplier No. 611F96BK
  • ONLINE EXCLUSIVE PRICE
    228.00 €
    142.00 € / Pack of 50
    Save 86.00 €
    37% Off
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Perform both fluorescence work and bioluminescence studies with 96-well Thermo Scientific™ Sterilin™ Black and White Microtiter™ plates.
Perform both fluorescence work and bioluminescence studies with 96-well Thermo Scientific™ Sterilin™ Black and White Microtiter™ plates. Supplied with no lid; if required, the 642000 lid is a suitable fit.
  • Black 96-well plates are ideal for fluorescence work where the black pigment prevents «cross-talk« between the wells — the low background fluorescence minimizes light scattering
  • White 96-well plates are ideal for bioluminescence studies where the pigment aids reflectivity and sensitivity
  • SBS compliant, ensuring suitable fit with all automation
  • Manufactured from virgin polystyrene
  • Supplied nonsterile, providing a medium binding surface
  • Chimney well design to help reduce contamination
Ordering Alerts:
Supplied without a lid; if required, the 642000 lid is a suitable fit.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Packaging 50 Cases, 5 Pk.
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
No. per Case 50
No. per Pack 5
Quantity 50/Pk.