missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Sterisart Bd Bottled Media: FTM, Wide-Mouth Bottle

Product Code. 15972710 Shop All Sartorius Products
Change view
Click to view available options
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15972710 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15972710 Supplier Sartorius Stedim Biotech™ Supplier No. BD257293
  • ONLINE EXCLUSIVE PRICE
    254.00 €
    238.23 € / Pack of 10
    Save 15.77 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterility testing fluid FTM is used to test biologics and for the enrichment and cultivation of anaerobes, aerobes, and microaerophiles.

  • Fluid thioglycollate medium
  • Liquid nutrient medium for the sterility testing of biologics and for the enrichment and cultivation of anaerobes, aerobes, and microaerophiles acc. to USP and EP.
  • Not under vacuum, but the headspace of the FTM is gassed with a stream of nitrogen to maintain anaerobiosis.
  • Contains a special quality of agar, resulting in a high clarity of the prepared medium..
TRUSTED_SUSTAINABILITY

Specifications

Cap Type Type 7 Closure, 45 mm OD
Media Type FTM (Fluid Thioglycollate Medium)
Sterility Sterile
No. per Pack 10
Packaging Type Wide-mouth bottle, individually double-packaged
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.