Learn More
STOML2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10348-100UL
 View more versions of this product
Product Code. 18018085
  • 450.00 € / 100µL
Description
The STOML2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human STOML2. This antibody recognizes human, mouse, rat antigen. The STOML2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
STOML2
1000 μg/mL
EPB72 like protein 2, HSPC108, SLP 2, SLP2, stomatin (EPB72) like 2, Stomatin like protein 2, STOML2
100 μL
30968
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa