Learn More
STOML2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10348-20UL
Description
The STOML2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human STOML2. This antibody recognizes human, mouse, rat antigen. The STOML2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| STOML2 | |
| 1000 μg/mL | |
| EPB72 like protein 2, HSPC108, SLP 2, SLP2, stomatin (EPB72) like 2, Stomatin like protein 2, STOML2 | |
| 20 μL | |
| 30968 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |