missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRA8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580082
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: A231 Cell, JURKAT Cell, HT1080 Cell, SCG Cell.
Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.
Specifications
| STRA8 | |
| Polyclonal | |
| Unconjugated | |
| STRA8 | |
| RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 346673 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q7Z7C7 | |
| STRA8 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human Stra8 (313-330aa DEEMIMLQCTETFDDEDL). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction