Learn More
Gosselin™ Straight Specimen Containers with Red Cap and Label
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand:  Gosselin™ TP52C-032
Code : NEW
Additional Details : Weight : 7.50000kg
 View more versions of this product
Product Code. 15468834
  • 184.67 € / Pack of 380
Estimated Shipment: 12-07-2024
to see stock.
Description
Description
  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
Specifications
Specifications
Specimen Container
Polypropylene
Sterile
Red
Clear
White label
380/Pk.
Yes
Straight
125 mL
Screw Cap
White label
380/Cs.