Learn More
Streptavidin Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB9020-SP
 View more versions of this product
Product Code. 18704469
  • 214.00 € / 25µg
Description
Streptavidin Monoclonal specifically detects Streptavidin in Multi-Species samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Streptavidin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Detects Streptavidin in Western blot.
Multi-species
Purified
Western Blot
1220C
Western Blot 0.5 ug/mL
SA V1; SA V2; Streptavidin V1; Streptavidin V2
Purified streptavidin from Streptomyces avidinii
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.