missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9020
This item is not returnable.
View return policy
Description
Streptavidin Monoclonal specifically detects Streptavidin in Multi-Species samples. It is validated for Western Blot.
Specifications
| Streptavidin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects Streptavidin in Western blot. | |
| Multi-species | |
| Purified |
| Western Blot | |
| 1220C | |
| Western Blot 0.5 ug/mL | |
| SA V1; SA V2; Streptavidin V1; Streptavidin V2 | |
| Purified streptavidin from Streptomyces avidinii | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction