missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptococcal Selective Agar C.O.B.A.

Product Code. 12997138
Change view
Click to view available options
Description:
Streptococcal Selective Agar C.O.B.A.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12997138 Streptococcal Selective Agar C.O.B.A. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12997138 Supplier Thermo Scientific™ Supplier No. PB0298A
  • ONLINE EXCLUSIVE PRICE
    72.85 €
    36.23 € / Pack of 10
    Save 36.62 €
    50% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate Streptococcus species with this ready-to-use medium which facilitetes uninhibited selective growth of streptococci species.

Selectively isolate Streptococcus species with Thermo Scientific™ Oxoid™ Streptococcal Selective Agar C.O.B.A. (Colistin sulphate Oxolinic acid Blood Agar) Thermo Scientific™ Oxoid™ Columbia Blood Agar forms the base for the medium to which antimicrobial agents and defibrinated horse blood are added to aid in the identification of hemolytic colonies.

The presence of selective agents colistin sulphate and oxolinic acid in the medium result in total inhibition of Gram-negative organisms and almost all non-streptococcal Gram-positive organisms, while having no inhibitory effects on Streptococcus species.

Use Streptococcal Selective Agar C.O.B.A. for selective isolation of streptococcal species with clearly defined hemolytic reactions.

  • Facilitates uninhibited selective growth of streptococci species, comparable to that of non-selective media.
  • Clearly defined hemolytic reactions with addition of defibrinated blood.
  • Ready-to-use convenience of prepared media.

Streptococci are commonly isolated from the upper respiratory tract. They are also often isolated from burns and other sites where frequently there is an abundance of competing organisms. In order to isolate streptococci, especially when present in small numbers, it is necessary to inhibit the competing flora without any adverse effect by the selective agents upon the Streptococcus species. The selective agents colistin sulphate and oxolinic acid have been found to have no inhibitory effect on Streptococcus species although amongst Group D organisms Enterococcus faecalis colonies are somewhat smaller. The combination of these two selective agents results in total inhibition of Gram-negative organisms and almost all non-streptococcal Gram-positive organisms. A very few staphylococci and coryneform organisms may grow with reduced colony size. The hemolytic reactions on media containing blood are clearly defined, and the colonial size and growth recovery of streptococcal groups A, B, C, D and G and S. pneumoniae are comparable to that on a non-selective medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 2°C to 10°C
Description Streptococcal Selective Agar C.O.B.A.
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.