missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Streptomycin sulfate

Product Code. 15593965 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15593965

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.

• It inhibits initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.

Application

• Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance

• This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture

• Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis

• This compound can be used as a protein synthesis inhibitor

• It acts against gram-negative and gram-positive bacteria • Streptomycin sulfate also shows a fungicide activity

• It can be used to remove nucleic acids from a biological extract

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Type Streptomycin Sulfate
Loss on Drying 7% max.
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 1457.4
Chemical Name or Material Streptomycin sulfate
compliance-icons compliance-icons
Product Identifier
  • Streptomycin Sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.