missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin Sulfate, Fisher BioReagents

Product Code. 10639933
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10639933 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10639933 Supplier Fisher BioReagents Supplier No. BP91050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Streptomycin sulfate can be used to prevent bacterial contamination of cell and tissue cultures
  •  Highly effective against gram-negative bacteria, and is often used in combination with penicillin
  • Acts by binding to the 30S subnit of bacterial ribosomes leading to inhibition of protein synthesis and death
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

Type Streptomycin Sulfate
Physical Form Solid
CAS 3810-74-0
pH 4.5 to 7.0
Loss on Drying < 5.0%
Packaging Poly Bottle
Potency Inclusive between 650μg/mg
Color White
Quantity 50 g
Molecular Formula C42H84N14O36S3
Linear Formula (C21H39N7O12)2·3H2SO4
Merck Index 15, 8956
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
Molecular Weight (g/mol) 1457.376
PubChem CID 124080941
Formula Weight 1457.58
Chemical Name or Material Streptomycin Sulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Streptomycin Sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.