missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Striatin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 539.00 €
Specifications
| Antigen | Striatin |
|---|---|
| Dilution | Western Blot 1:100-1:500, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18400091
|
Novus Biologicals
NBP1-80762 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18449430
|
Novus Biologicals
NBP1-80762-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Striatin Polyclonal antibody specifically detects Striatin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Striatin | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 6801 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 1:100-1:500, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC125642, SG2NA, striatin, striatin, calmodulin binding protein, striatin, calmodulin-binding protein | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RALLGFSSDVTDREDDKNQDSVVNGTEAEVKETAMIAKSELTDSASVLDNFKFLESAAADFSDEDEDDDVDGREKSVIDTSTIVRKKALPDSGEDRDTKEALKEFDFLVTSEEGDNESRSAGDGTDWEKEDQC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title