missing translation for 'onlineSavingsMsg'
Learn More

Strontium for AAS 1g/L HCl matrix, Fisher chemical™
BIOPHARMA

Product Code. 12953225 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12953225 500 mL Bottle 500mL
12943225 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12953225 Supplier Fisher Chemical Supplier No. B 056 500
  • ONLINE EXCLUSIVE PRICE
    60.00 €
    46.17 € / 500mL
    Save 13.83 €
    23% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Identificadores de productos químicos

CAS 7440-24-6
Molecular Formula Sr
Molecular Weight (g/mol) 87.62
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
PubChem CID 5359327
ChEBI CHEBI:33324
IUPAC Name strontium
SMILES [Sr]

Especificaciones

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix 2% HCl
Quantity 500 mL
Chemical Name or Material Strontium
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.