missing translation for 'onlineSavingsMsg'
Learn More

Strontium Ranelate, MedChemExpress

Product Code. 16461846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
16461846 500 mg 500mg
16407286 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 16461846

Brand: Medchem Express HY17397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Strontium Ranelate (S12911) is an antiosteoporotic agent that acts by reducing bone resorption and promoting bone formation, thereby inducing a positive bone balance. Strontium Ranelate can also activate the calcium-sensing receptor (CaSR) in non skeletal cells, resulting in the activation of inositol 1, 4, 5-triphosphate production and mitogen-activated protein kinase signaling.

Strontium Ranelate (S12911) is an antiosteoporotic agent that acts by reducing bone resorption and promoting bone formation, thereby inducing a positive bone balance. Strontium Ranelate can also activate the calcium-sensing receptor (CaSR) in non skeletal cells, resulting in the activation of inositol 1, 4, 5-triphosphate production and mitogen-activated protein kinase signaling.

Specifications

Chemical Name or Material Strontium Ranelate
Physical Form Solid
CAS 135459-87-9
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H6N2O8SSr2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Distrontium renelate S12911
Solubility Information 0.1 M HCL : 17.5 mg/mL (34.08 mM; ultrasonic and adjust pH to 3 with HCl) ∣H2O : 2 mg/mL (3.89 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(O[Sr]OC(C1)=O)C2=C1C(C#N)=C(N3CC(O[Sr]OC(C3)=O)=O)S2
Molecular Weight (g/mol) 513.49
Formula Weight 513.49
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.