missing translation for 'onlineSavingsMsg'
Learn More

Strontium Standard for ICP/MS, 2% HNO3, SPEX CertiPrep™

Product Code. 15236006 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Pakningsstørrelse:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
15236006 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15236006 Leverandør SPEX Certiprep Leverandørnr. CLSR21BY
  • ONLINE EXCLUSIVE PRICE
    129.69 €
    112.73 € / 125mL
    Save 16.96 €
    13% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Indeholder

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sr1μg/mL

Certificeringer

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Overholdelse

GHS/OSHA Complaint

Tekniske data

Packaging Triple-leached LDPE bottle
CAS 7440-24-6, 7697-37-2, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 2.0, 0.0, 98.0
Density 1.01006 g/cm3 at 20°C
Physical Form Liquid
Molecular Formula Sr
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
SMILES [Sr]
IUPAC Name strontium
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 87.62
PubChem CID 5359327
Quantity 125 mL
ChEBI CHEBI:33324
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Strontium Standard, ICP/MS
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 1
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Sikkerhedssætning
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplerende oplysninger
  • MIXTURE LIST-Contain: nitric acid

Product intended for laboratory use only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.