missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STT3B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93315-0.1ml
This item is not returnable.
View return policy
Description
STT3B Polyclonal antibody specifically detects STT3B in Human, Mouse samples. It is validated for Western Blot
Specifications
| STT3B | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| FLJ90106, homolog of yeast STT3, Oligosaccharyl transferase subunit STT3B, SIMPEC 2.4.1.119, source of immunodominant MHC associated peptides, Source of immunodominant MHC-associated peptides homolog, STT3, subunit of the oligosaccharyltransferase complex, homolog B (S.cerevisiae), STT3-Bdolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human STT3B (NP_849193.1). MAEPSAPESKHKSSLNSSPWSGLMALGNSRHGHHGPGAQCAHKAAGGAAPPKPAPAGLSGGLSQPAGWQSLLSFTILFLAWLAGFSSRLFAVIRFESIIH | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 201595 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction