Learn More
Cole-Parmer™ Dual Tube Holder Disc Adapter for Cole-Parmer™ TR-200 Series Tube Rotators
Description
- Choose the tube holder(s) that accommodates your tube size(s).
- Fits two tube holders to Cole-Parmer™ TR-200 series tube rotator simultaneously.
- 1 year warranty.
Specifications
Specifications
| Product Type | Adapter |
| For Use With (Equipment) | Stuart™ SB2/SB3 Tube Rotator Holders |