Learn More
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators
Description
- Choose the tube holder(s) that accommodates your tube size(s).
Specifications
Specifications
| Holds | Test Tubes, Blood Tubes |
| Tube Size | 9 to 20 mm |
| Product Type | Tube Holder |
| For Use With (Equipment) | For 20 Glaser |
| No. of Positions | 20 Tubes |