Learn More
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators
Product Code. 12027144
Change view
Click to view available options
For Use With (Equipment):
For 12 Tubes
For 20 Glaser
For 30 Tubes, 26 mm dia.
For 40 Microcentrifuge Tubes
For 63 Tubes, 12 mm dia.
For 63 Tubes, 16 mm dia.
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12027144 For 12 Tubes Each
11416558 For 20 Glaser Each
11446558 For 30 Tubes, 26 mm dia. Each
11406558 For 40 Microcentrifuge Tubes Each
12037144 For 63 Tubes, 12 mm dia. Each
11436558 For 63 Tubes, 16 mm dia. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12027144 Supplier Cole-Parmer™ Supplier No. 0007713
Only null left
Add to basket
Add to basket
Quickly and easily replace tube holder without tools using these Tube Holder Discs for Cole-Parmer™ TR-200 Series Tube Rotators.
  • Choose the tube holder(s) that accommodates your tube size(s).
Specifications
Holds Centrifuge tubes
Tube Capacity 12 x 50 mL
Product Type Tube Holder
For Use With (Equipment) For 12 Tubes
No. of Positions 12 Tubes