Learn More
Cole-Parmer™ Replacement Clip for Cole-Parmer™ TR-200 Series Tube Rotators
Product Code. 12869435
Change view
Click to view available options
For Use With (Equipment):
SB3/1 Tube Holder
Unit Size:
Pack of 20
Product Code. For Use With (Equipment) Unit Size
12869435 SB3/1 Tube Holder Pack of 20
1 options
Product Code. 12869435 Supplier Cole-Parmer™ Supplier No. 0007725
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
In Stock
Add to basket
Edge
Add to basket
Replacement Clip for Cole-Parmer™ TR-200 Series Tube Rotators replaces broken or weakened micro tube clips for the Stuart SB3/1 Tube Holder. Holds 10 to 11.5 mL micro tubes in place.
  • Choose the tube holder(s) that accommodates your tube size(s).
  • Spare clips for microcentrifuge tube holder disc 00077-11.
  • 1 year warranty.
Specifications
Product Type Spare Clip
For Use With (Equipment) SB3/1 Tube Holder