missing translation for 'onlineSavingsMsg'
Learn More

Succinate, 0.2M buffer soln., pH 5.0, Thermo Scientific™

Product Code. 15446019
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446019 500 mL 500mL
15436019 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446019 Supplier Thermo Scientific Alfa Aesar Supplier No. J63863.AP
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00002789
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Concentration 0.2 M
pH 5.0
Physical Form Liquid
Quantity 500 mL
Odor Odorless
Chemical Name or Material Succinate buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.