missing translation for 'onlineSavingsMsg'
Learn More

Succinyl phosphonate trisodium salt, MedChemExpress

Product Code. 16478007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478007 5 mg 5mg
16415298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478007 Supplier Medchem Express Supplier No. HY12688A5mg
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Succinyl phosphonate trisodium salt is an α-ketoglutarate dehydrogenase (KGDHC) inhibitor, effective inhibits (KGDHC) in muscle, bacterial, brain, and cultured human fibroblasts.

Succinyl phosphonate trisodium salt is an 2-oxoglutarate dehydrogenase (OGDH) inhibitor, impairs viability of cancer cells in a cell-specific metabolism-dependent manner. Succinyl phosphonate trisodium salt inhibits the glutamate-induced ROS production in glutamate-stimulated hippocampal neurons in situ.

Specifications

Chemical Name or Material Succinyl phosphonate trisodium salt
Physical Form Solid
CAS 864167-45-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C4H4Na3O6P
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information H2O : 150 mg/mL (604.81 mM; Need ultrasonic)
SMILES O=C(O[Na])CCC(P(O[Na])(O[Na])=O)=O
Molecular Weight (g/mol) 248.01
Formula Weight 248.01
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.